Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTTGATAGAAGACAGTCAATATC[A/G]TGAAGGATTTCAGAGGTTTTTTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf34 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C5orf34 - chromosome 5 open reading frame 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198566.3 | 2224 | Silent Mutation | CAC,CAT | H,H 627 | NP_940968.1 | |
XM_011514038.2 | 2224 | UTR 3 | XP_011512340.1 | |||
XM_017009445.1 | 2224 | Silent Mutation | CAC,CAT | H,H 513 | XP_016864934.1 | |
XM_017009446.1 | 2224 | Silent Mutation | CAC,CAT | H,H 513 | XP_016864935.1 |
TMEM267 - transmembrane protein 267 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |