Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGAGGGCCTCAAGCCCACACTG[A/G]CTTATTTGAAATCCATGTTTAGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS7BP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RGS7BP - regulator of G-protein signaling 7 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029875.2 | 1205 | Intron | NP_001025046.1 | |||
NM_001271890.1 | 1205 | Intron | NP_001258819.1 | |||
NM_001271891.1 | 1205 | Intron | NP_001258820.1 | |||
XM_005248502.3 | 1205 | UTR 5 | XP_005248559.1 |