Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGTGTACGGGGCTGGAGGGGCC[A/G]CCTAGAGATTGCCAGCTTCCTACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134935 MIM: 605308 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGFR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGFR4 - fibroblast growth factor receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291980.1 | 421 | Missense Mutation | CAC,CGC | H,R 85 | NP_001278909.1 | |
NM_002011.4 | 421 | Missense Mutation | CAC,CGC | H,R 85 | NP_002002.3 | |
NM_022963.3 | 421 | Missense Mutation | CAC,CGC | H,R 85 | NP_075252.2 | |
NM_213647.2 | 421 | Missense Mutation | CAC,CGC | H,R 85 | NP_998812.1 | |
XM_005265838.3 | 421 | Missense Mutation | CAC,CGC | H,R 85 | XP_005265895.1 | |
XM_011534464.2 | 421 | Missense Mutation | CAC,CGC | H,R 116 | XP_011532766.1 | |
XM_011534465.2 | 421 | UTR 5 | XP_011532767.1 |
ZNF346 - zinc finger protein 346 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |