Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAACAATTATGAATGAGGCATTTA[A/G]TAAAAACATTTTATTAATTCCTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603335 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAH5 - dynein axonemal heavy chain 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001369.2 | 10948 | UTR 3 | NP_001360.1 | |||
XM_005248262.3 | 10948 | UTR 3 | XP_005248319.2 | |||
XM_017009177.1 | 10948 | UTR 3 | XP_016864666.1 | |||
XM_017009178.1 | 10948 | UTR 3 | XP_016864667.1 | |||
XM_017009179.1 | 10948 | UTR 3 | XP_016864668.1 | |||
XM_017009180.1 | 10948 | Intron | XP_016864669.1 | |||
XM_017009181.1 | 10948 | Intron | XP_016864670.1 | |||
XM_017009182.1 | 10948 | Intron | XP_016864671.1 | |||
XM_017009183.1 | 10948 | Intron | XP_016864672.1 | |||
XM_017009184.1 | 10948 | Intron | XP_016864673.1 | |||
XM_017009185.1 | 10948 | UTR 3 | XP_016864674.1 | |||
XM_017009186.1 | 10948 | UTR 3 | XP_016864675.1 | |||
XM_017009187.1 | 10948 | Intron | XP_016864676.1 | |||
XM_017009188.1 | 10948 | UTR 3 | XP_016864677.1 |