Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCTCTGCATCAGCCTCATCAGT[C/T]CTGTCATAGGTCAGGAGGTCTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602121 MIM: 605166 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIAPH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DIAPH1 - diaphanous related formin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HDAC3 - histone deacetylase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003883.3 | 1233 | Silent Mutation | NP_003874.2 | |||
XM_011537697.2 | 1233 | Missense Mutation | XP_011535999.1 |