Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAACAGTAAGTATTTGGTAACA[A/G]TGATTAACCATGAAAGATCAGTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606141 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RANBP17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RANBP17 - RAN binding protein 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022897.4 | Intron | NP_075048.1 | ||||
XM_011534627.1 | Intron | XP_011532929.1 | ||||
XM_011534631.2 | Intron | XP_011532933.1 | ||||
XM_011534636.2 | Intron | XP_011532938.1 | ||||
XM_011534637.2 | Intron | XP_011532939.1 | ||||
XM_017009736.1 | Intron | XP_016865225.1 | ||||
XM_017009737.1 | Intron | XP_016865226.1 | ||||
XM_017009738.1 | Intron | XP_016865227.1 | ||||
XM_017009739.1 | Intron | XP_016865228.1 | ||||
XM_017009740.1 | Intron | XP_016865229.1 | ||||
XM_017009741.1 | Intron | XP_016865230.1 | ||||
XM_017009742.1 | Intron | XP_016865231.1 | ||||
XM_017009743.1 | Intron | XP_016865232.1 | ||||
XM_017009744.1 | Intron | XP_016865233.1 | ||||
XM_017009745.1 | Intron | XP_016865234.1 | ||||
XM_017009746.1 | Intron | XP_016865235.1 | ||||
XM_017009747.1 | Intron | XP_016865236.1 |