Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCTCTGTCCGTGCAGGTCCATC[C/T]ACATCGTGGCGCTGGGGAACGAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607353 MIM: 608039 | ||||||||||||||||||||
Literature Links: |
RHOBTB3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RHOBTB3 - Rho related BTB domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014899.3 | 547 | Missense Mutation | CAC,TAC | H,Y 4 | NP_055714.3 | |
XM_011543279.2 | 547 | Missense Mutation | CAC,TAC | H,Y 4 | XP_011541581.1 | |
XM_017009236.1 | 547 | UTR 5 | XP_016864725.1 | |||
XM_017009237.1 | 547 | UTR 5 | XP_016864726.1 |
SPATA9 - spermatogenesis associated 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031952.3 | 547 | Intron | NP_114158.2 | |||
XM_011543664.1 | 547 | Intron | XP_011541966.1 | |||
XM_011543666.1 | 547 | Intron | XP_011541968.1 | |||
XM_011543667.2 | 547 | Intron | XP_011541969.1 | |||
XM_017009948.1 | 547 | Intron | XP_016865437.1 | |||
XM_017009949.1 | 547 | Intron | XP_016865438.1 | |||
XM_017009950.1 | 547 | Intron | XP_016865439.1 | |||
XM_017009951.1 | 547 | Intron | XP_016865440.1 |