Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTTCTTGCCCAAGAGAGGTCTTC[C/T]TTCCATATTTCAGGGTTCATTGGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601586 | ||||||||||||||||||||
Literature Links: |
PTGER4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTGER4 - prostaglandin E receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000958.2 | 1768 | Intron | NP_000949.1 | |||
XM_017009656.1 | 1768 | Intron | XP_016865145.1 | |||
XM_017009657.1 | 1768 | Intron | XP_016865146.1 | |||
XM_017009658.1 | 1768 | Intron | XP_016865147.1 | |||
XM_017009659.1 | 1768 | Intron | XP_016865148.1 |
TTC33 - tetratricopeptide repeat domain 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012382.2 | 1768 | Silent Mutation | AAA,AAG | K,K 165 | NP_036514.1 | |
XM_011514003.2 | 1768 | Silent Mutation | AAA,AAG | K,K 165 | XP_011512305.1 |