Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACAATAAATGTGAAGACATGTTT[C/G]TCTGGGATGTGGTTTTTTTTTTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615189 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD55 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD55 - ankyrin repeat domain 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024669.2 | 1734 | UTR 3 | NP_078945.2 | |||
XM_011543646.1 | 1734 | UTR 3 | XP_011541948.1 | |||
XM_017009852.1 | 1734 | UTR 3 | XP_016865341.1 | |||
XM_017009853.1 | 1734 | UTR 3 | XP_016865342.1 | |||
XM_017009854.1 | 1734 | UTR 3 | XP_016865343.1 |