Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCGCCGCCCGTGCCGCCGCCATG[G/T]TGATGTCCCAGGGCACCTACACGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126660 | ||||||||||||||||||||
Literature Links: |
DBN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DBN1 - drebrin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR7 - proline rich 7 (synaptic) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174101.1 | 1202 | Missense Mutation | GTG,TTG | V,L 2 | NP_001167572.1 | |
NM_001174102.1 | 1202 | Missense Mutation | GTG,TTG | V,L 2 | NP_001167573.1 | |
NM_030567.4 | 1202 | Missense Mutation | GTG,TTG | V,L 2 | NP_085044.2 | |
XM_011534662.1 | 1202 | Missense Mutation | GTG,TTG | V,L 2 | XP_011532964.1 | |
XM_011534663.2 | 1202 | Missense Mutation | GTG,TTG | V,L 2 | XP_011532965.1 | |
XM_017009896.1 | 1202 | Missense Mutation | GTG,TTG | V,L 2 | XP_016865385.1 | |
XM_017009897.1 | 1202 | Missense Mutation | GTG,TTG | V,L 2 | XP_016865386.1 |
PRR7-AS1 - PRR7 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |