Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACTTACATTTTTATGTACAGGAA[A/C]AGGAATCCTCAGAGTCTGGAGAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147850 | ||||||||||||||||||||
Literature Links: |
IL5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IL5 - interleukin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000879.2 | 204 | Missense Mutation | NP_000870.1 | |||
XM_005271988.3 | 204 | Missense Mutation | XP_005272045.1 | |||
XM_011543373.2 | 204 | Missense Mutation | XP_011541675.1 | |||
XM_011543374.2 | 204 | Missense Mutation | XP_011541676.1 |