Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTGCAGAAATTTTATCCAACTT[C/T]GTTTGGAAGGTATGTTGACTTTTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CWC27 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CWC27 - CWC27 spliceosome associated protein homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297644.1 | 359 | Missense Mutation | CGT,TGT | R,C 44 | NP_001284573.1 | |
NM_001297645.1 | 359 | Missense Mutation | CGT,TGT | R,C 44 | NP_001284574.1 | |
NM_001318000.1 | 359 | Missense Mutation | CGT,TGT | R,C 44 | NP_001304929.1 | |
NM_005869.3 | 359 | Missense Mutation | CGT,TGT | R,C 44 | NP_005860.2 | |
XM_011543095.2 | 359 | Missense Mutation | CGT,TGT | R,C 44 | XP_011541397.2 | |
XM_011543096.1 | 359 | Missense Mutation | CGT,TGT | R,C 44 | XP_011541398.1 | |
XM_017008943.1 | 359 | Missense Mutation | CGT,TGT | R,C 44 | XP_016864432.1 |
SREK1IP1 - SREK1 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |