Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGACGAAGCAGGTGTCATGCCA[A/G]CTGAAGCCCAGGGCCTCCAGGAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126660 MIM: 605903 | ||||||||||||||||||||
Literature Links: |
DBN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DBN1 - drebrin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDLIM7 - PDZ and LIM domain 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005451.4 | 1349 | Silent Mutation | AGC,AGT | S,S 420 | NP_005442.2 | |
NM_203352.2 | 1349 | Silent Mutation | AGC,AGT | S,S 386 | NP_976227.1 | |
NM_213636.2 | 1349 | Intron | NP_998801.1 | |||
XM_011534698.2 | 1349 | Silent Mutation | AGC,AGT | S,S 327 | XP_011533000.1 | |
XM_011534699.2 | 1349 | Silent Mutation | AGC,AGT | S,S 272 | XP_011533001.1 |