Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAAGATAACTATAGAATGAAAA[A/G]TTATAAGAATAAAGCCCTAAATCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604545 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KPNA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KPNA5 - karyopherin subunit alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002269.2 | 159 | Missense Mutation | AAT,AGT | N,S 16 | NP_002260.2 | |
XM_011535805.1 | 159 | Missense Mutation | AAT,AGT | N,S 36 | XP_011534107.1 | |
XM_017010838.1 | 159 | Missense Mutation | AAT,AGT | N,S 36 | XP_016866327.1 | |
XM_017010839.1 | 159 | Missense Mutation | AAT,AGT | N,S 16 | XP_016866328.1 | |
XM_017010840.1 | 159 | Missense Mutation | AAT,AGT | N,S 13 | XP_016866329.1 | |
XM_017010841.1 | 159 | Intron | XP_016866330.1 | |||
XM_017010842.1 | 159 | UTR 5 | XP_016866331.1 | |||
XM_017010843.1 | 159 | UTR 5 | XP_016866332.1 |