Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGCCCTCGGTGGCCCCCTCCCCC[A/G]TGGCCAGGACCATCATGGCCACGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603771 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R10 - protein phosphatase 1 regulatory subunit 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002714.3 | 3270 | Silent Mutation | CAC,CAT | H,H 888 | NP_002705.2 | |
XM_006715130.2 | 3270 | Silent Mutation | CAC,CAT | H,H 888 | XP_006715193.1 | |
XM_011514722.1 | 3270 | Silent Mutation | CAC,CAT | H,H 888 | XP_011513024.1 | |
XM_017010995.1 | 3270 | Silent Mutation | CAC,CAT | H,H 888 | XP_016866484.1 |