Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGGCGTTAGGGGCGTCCTCTGGA[A/G]GCAGGGGCGCCGGCACACCCTCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606998 MIM: 602996 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLOT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLOT1 - flotillin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HCG20 - HLA complex group 20 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IER3 - immediate early response 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003897.3 | 361 | Missense Mutation | CCT,CTT | P,L 110 | NP_003888.2 |