Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTGACCAAGGCCTGTCGGTCAC[A/G]GGGCAGAGAGAGCCGGTGCAGAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603871 MIM: 123834 | ||||||||||||||||||||
Literature Links: |
BYSL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BYSL - bystin like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCND3 - cyclin D3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136017.3 | 497 | Missense Mutation | CGT,TGT | R,C 91 | NP_001129489.1 | |
NM_001136125.2 | 497 | Missense Mutation | CGT,TGT | R,C 100 | NP_001129597.1 | |
NM_001136126.2 | 497 | UTR 5 | NP_001129598.1 | |||
NM_001287427.1 | 497 | Missense Mutation | CGT,TGT | R,C 122 | NP_001274356.1 | |
NM_001287434.1 | 497 | UTR 5 | NP_001274363.1 | |||
NM_001760.4 | 497 | Missense Mutation | CGT,TGT | R,C 172 | NP_001751.1 | |
XM_011514971.1 | 497 | Missense Mutation | CGT,TGT | R,C 172 | XP_011513273.1 | |
XM_011514972.1 | 497 | Missense Mutation | CGT,TGT | R,C 91 | XP_011513274.1 |