Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCTGGGTGACCCGGCCTTGGAAG[C/T]TGAGGGTGTAGGAGCCACTGTCATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603170 MIM: 602280 | ||||||||||||||||||||
Literature Links: |
TEAD3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TEAD3 - TEA domain transcription factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TULP1 - tubby like protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289395.1 | 1477 | Missense Mutation | AAC,AGC | N,S 425 | NP_001276324.1 | |
NM_003322.4 | 1477 | Missense Mutation | AAC,AGC | N,S 478 | NP_003313.3 |