Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601417 MIM: 611500 MIM: 602045 MIM: 180246 MIM: 601416 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD17B8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSD17B8 - hydroxysteroid 17-beta dehydrogenase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014234.4 | 66 | Missense Mutation | GCA,TCA | A,S 12 | NP_055049.1 |
MIR219A1 - microRNA 219a-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RING1 - ring finger protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RXRB - retinoid X receptor beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC39A7 - solute carrier family 39 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077516.1 | 66 | Intron | NP_001070984.1 | |||
NM_001288777.1 | 66 | Intron | NP_001275706.1 | |||
NM_006979.2 | 66 | Intron | NP_008910.2 |