Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTGAGAGCTATTCTTGAGAGCT[G/T]GTGCTGCCTAAAAGGTCTCAATTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601594 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JARID2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JARID2 - jumonji and AT-rich interaction domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267040.1 | Intron | NP_001253969.1 | ||||
NM_004973.3 | Intron | NP_004964.2 | ||||
XM_005249089.3 | Intron | XP_005249146.1 | ||||
XM_011514579.2 | Intron | XP_011512881.1 | ||||
XM_011514584.2 | Intron | XP_011512886.1 | ||||
XM_017010833.1 | Intron | XP_016866322.1 | ||||
XM_017010834.1 | Intron | XP_016866323.1 | ||||
XM_017010835.1 | Intron | XP_016866324.1 |
JARID2-AS1 - JARID2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |