Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGTTAAACATATACTAGATCGC[A/G]ATTTTTAATCTCTGAGCCTGTGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603473 | ||||||||||||||||||||
Literature Links: |
LATS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LATS1 - large tumor suppressor kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270519.1 | 3782 | Intron | NP_001257448.1 | |||
NM_004690.3 | 3782 | Missense Mutation | CGC,TGC | R,C 1125 | NP_004681.1 | |
XM_006715603.3 | 3782 | Intron | XP_006715666.1 | |||
XM_011536252.2 | 3782 | Intron | XP_011534554.1 | |||
XM_017011474.1 | 3782 | Missense Mutation | CGC,TGC | R,C 1158 | XP_016866963.1 | |
XM_017011475.1 | 3782 | Missense Mutation | CGC,TGC | R,C 1158 | XP_016866964.1 | |
XM_017011476.1 | 3782 | Missense Mutation | CGC,TGC | R,C 1053 | XP_016866965.1 | |
XM_017011477.1 | 3782 | Intron | XP_016866966.1 | |||
XM_017011478.1 | 3782 | Missense Mutation | CGC,TGC | R,C 1020 | XP_016866967.1 | |
XM_017011479.1 | 3782 | UTR 3 | XP_016866968.1 | |||
XM_017011480.1 | 3782 | Missense Mutation | CGC,TGC | R,C 878 | XP_016866969.1 | |
XM_017011481.1 | 3782 | Missense Mutation | CGC,TGC | R,C 845 | XP_016866970.1 | |
XM_017011482.1 | 3782 | Intron | XP_016866971.1 |