Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAACAACTGCTGTTCCGATAGA[A/G]GAACCCAGTGCAGCTGCCTGCATGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180297 | ||||||||||||||||||||
Literature Links: |
RHAG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RHAG - Rh-associated glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000324.2 | 1163 | Silent Mutation | TCC,TCT | S,S 367 | NP_000315.2 | |
XM_011514788.1 | 1163 | Intron | XP_011513090.1 |