Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAGGTTCTTCGTGTTCAATGTTA[C/T]GTTCAGTACAAAACCTGATGTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609206 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEF1E1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EEF1E1 - eukaryotic translation elongation factor 1 epsilon 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135650.1 | Intron | NP_001129122.1 | ||||
NM_004280.4 | Intron | NP_004271.1 |
EEF1E1-BLOC1S5 - EEF1E1-BLOC1S5 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCARNA27 - small Cajal body-specific RNA 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |