Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGTCCAGTTCAACTGCATCGGCCG[C/T]GCGCGCAGCTGGAACGAGAAGCAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607977 MIM: 611438 | ||||||||||||||||||||
Literature Links: |
HECA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HECA - hdc homolog, cell cycle regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016217.2 | 735 | Silent Mutation | CGC,CGT | R,R 150 | NP_057301.1 |
TXLNB - taxilin beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153235.3 | 735 | Intron | NP_694967.3 | |||
XM_005266836.1 | 735 | Intron | XP_005266893.1 | |||
XM_011535505.2 | 735 | Intron | XP_011533807.1 | |||
XM_011535506.2 | 735 | Intron | XP_011533808.1 | |||
XM_017010322.1 | 735 | Intron | XP_016865811.1 |