Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTATACTAAATGTTAGTAACTGT[A/G]GTACTTCATGATGATCAGGAACCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TULP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TULP4 - tubby like protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007466.2 | Intron | NP_001007467.1 | ||||
NM_020245.4 | Intron | NP_064630.2 | ||||
XM_011535946.1 | Intron | XP_011534248.1 | ||||
XM_017011063.1 | Intron | XP_016866552.1 | ||||
XM_017011064.1 | Intron | XP_016866553.1 | ||||
XM_017011065.1 | Intron | XP_016866554.1 | ||||
XM_017011066.1 | Intron | XP_016866555.1 | ||||
XM_017011067.1 | Intron | XP_016866556.1 | ||||
XM_017011068.1 | Intron | XP_016866557.1 | ||||
XM_017011069.1 | Intron | XP_016866558.1 | ||||
XM_017011070.1 | Intron | XP_016866559.1 | ||||
XM_017011071.1 | Intron | XP_016866560.1 |