Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCACCCTGCACTTACCTGCATG[C/G]TGCCGCTTCTTCCTGCCCACAGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606627 MIM: 603707 | ||||||||||||||||||||
Literature Links: |
DAAM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DAAM2 - dishevelled associated activator of morphogenesis 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOCS1 - molybdenum cofactor synthesis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001075098.3 | 1140 | Missense Mutation | CAC,CAG | H,Q 365 | NP_001068566.1 | |
NM_005943.5 | 1140 | Missense Mutation | CAC,CAG | H,Q 365 | NP_005934.2 | |
XM_011514632.2 | 1140 | Missense Mutation | CAC,CAG | H,Q 365 | XP_011512934.1 | |
XM_011514633.2 | 1140 | Missense Mutation | CAC,CAG | H,Q 365 | XP_011512935.1 | |
XM_011514634.2 | 1140 | Missense Mutation | CAC,CAG | H,Q 278 | XP_011512936.1 | |
XM_017010888.1 | 1140 | Missense Mutation | CAC,CAG | H,Q 278 | XP_016866377.1 |