Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGCCCCAGGAGGCACCTGCTCC[C/T]GAGGCCTCTGCTGTGGGAGGGCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602623 | ||||||||||||||||||||
Literature Links: |
ARMC12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARMC12 - armadillo repeat containing 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286574.1 | 293 | Intron | NP_001273503.1 | |||
NM_001286576.1 | 293 | Intron | NP_001273505.1 | |||
NM_145028.4 | 293 | Silent Mutation | CCC,CCT | P,P 72 | NP_659465.2 | |
XM_011514381.2 | 293 | Intron | XP_011512683.1 | |||
XM_011514382.2 | 293 | Intron | XP_011512684.1 | |||
XM_017010435.1 | 293 | Silent Mutation | CCC,CCT | P,P 72 | XP_016865924.1 |
FKBP5 - FK506 binding protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC285847 - uncharacterized LOC285847 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |