Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGCAGATGATGCCCGCCGCGCA[C/T]ATCAACTCGCACCTGGACCGCTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608196 | ||||||||||||||||||||
Literature Links: |
MYLK4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYLK4 - myosin light chain kinase family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005249078.3 | 284 | Intron | XP_005249135.2 | |||
XM_005249079.2 | 284 | Intron | XP_005249136.1 | |||
XM_006715082.3 | 284 | Intron | XP_006715145.1 | |||
XM_011514567.2 | 284 | Intron | XP_011512869.1 | |||
XM_017010819.1 | 284 | Intron | XP_016866308.1 |
WRNIP1 - Werner helicase interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020135.2 | 284 | Silent Mutation | CAC,CAT | H,H 31 | NP_064520.2 | |
NM_130395.2 | 284 | Silent Mutation | CAC,CAT | H,H 31 | NP_569079.1 | |
XM_005249232.3 | 284 | Silent Mutation | CAC,CAT | H,H 31 | XP_005249289.1 | |
XM_017011060.1 | 284 | Silent Mutation | CAC,CAT | H,H 31 | XP_016866549.1 |