Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCGGGCGGGCGACGCTGGTCCAG[A/G]ACGGCATCGCCAAGGGGCGCGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTN1A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTN1A1 - butyrophilin subfamily 1 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001732.2 | 318 | Missense Mutation | AAC,GAC | N,D 100 | NP_001723.2 | |
XM_005249340.3 | 318 | Missense Mutation | AAC,GAC | N,D 200 | XP_005249397.2 |