Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAAGCCCAGAAGAGCCACCACACG[A/G]CCCCAATTGATGCCACTCTCAAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600516 MIM: 609495 | ||||||||||||||||||||
Literature Links: |
BAK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BAK1 - BCL2 antagonist/killer 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001188.3 | Intron | NP_001179.1 | ||||
XM_011514779.2 | Intron | XP_011513081.1 | ||||
XM_011514780.1 | Intron | XP_011513082.1 |
GGNBP1 - gametogenetin binding protein 1 (pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |