Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAATTCTTCCTCCCCTAAGGACTT[C/T]CTCATTGCTGTTTCTTCATCTTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615876 | ||||||||||||||||||||
Literature Links: |
RSPH3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RSPH3 - radial spoke 3 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031924.4 | 1164 | Silent Mutation | AGA,AGG | R,R 551 | NP_114130.3 | |
XM_005267153.4 | 1164 | Silent Mutation | AGA,AGG | R,R 455 | XP_005267210.1 | |
XM_017011347.1 | 1164 | Silent Mutation | AGA,AGG | R,R 279 | XP_016866836.1 |