Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCGAGCAAAAGTCAAATGATGC[C/T]TCTGAGGCGGAGCACCAGAGAGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 143010 | ||||||||||||||||||||
Literature Links: |
HLA-E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HLA-E - major histocompatibility complex, class I, E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005516.5 | 651 | Silent Mutation | GCC,GCT | A,A 171 | NP_005507.3 | |
XM_017010807.1 | 651 | Silent Mutation | GCC,GCT | A,A 212 | XP_016866296.1 | |
XM_017010808.1 | 651 | Silent Mutation | GCC,GCT | A,A 212 | XP_016866297.1 | |
XM_017010809.1 | 651 | Silent Mutation | GCC,GCT | A,A 171 | XP_016866298.1 |