Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTTCACAAATTTTAGAAACTCT[C/T]TATCTGTTAAAGAAATTATATTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159559 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFDN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AFDN - afadin, adherens junction formation factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040000.2 | Intron | NP_001035089.1 | ||||
NM_001207008.1 | Intron | NP_001193937.1 | ||||
NM_001291964.1 | Intron | NP_001278893.1 | ||||
XM_005266996.3 | Intron | XP_005267053.1 | ||||
XM_005266997.2 | Intron | XP_005267054.1 | ||||
XM_006715491.2 | Intron | XP_006715554.1 | ||||
XM_006715494.3 | Intron | XP_006715557.1 | ||||
XM_011535842.2 | Intron | XP_011534144.1 | ||||
XM_017010878.1 | Intron | XP_016866367.1 | ||||
XM_017010879.1 | Intron | XP_016866368.1 | ||||
XM_017010880.1 | Intron | XP_016866369.1 | ||||
XM_017010881.1 | Intron | XP_016866370.1 | ||||
XM_017010882.1 | Intron | XP_016866371.1 | ||||
XM_017010883.1 | Intron | XP_016866372.1 | ||||
XM_017010884.1 | Intron | XP_016866373.1 | ||||
XM_017010885.1 | Intron | XP_016866374.1 | ||||
XM_017010886.1 | Intron | XP_016866375.1 | ||||
XM_017010887.1 | Intron | XP_016866376.1 |