Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGTGAGCGATTCCATGCCTGTGC[A/G]GGGGTTGTAGCTCTTCTGCTTACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603405 | ||||||||||||||||||||
Literature Links: |
C6orf136 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C6orf136 - chromosome 6 open reading frame 136 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DHX16 - DEAH-box helicase 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164239.1 | 2527 | Missense Mutation | CGC,TGC | R,C 643 | NP_001157711.1 | |
NM_003587.4 | 2527 | Missense Mutation | CGC,TGC | R,C 703 | NP_003578.2 | |
XM_011514938.2 | 2527 | Missense Mutation | CGC,TGC | R,C 404 | XP_011513240.1 | |
XM_011514939.2 | 2527 | Missense Mutation | CGC,TGC | R,C 404 | XP_011513241.1 | |
XM_011514940.2 | 2527 | Missense Mutation | CGC,TGC | R,C 222 | XP_011513242.1 | |
XM_011514941.2 | 2527 | Missense Mutation | CGC,TGC | R,C 186 | XP_011513243.1 |