Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTTCCAACGCAGTGTCTCAAAT[A/G]TAGGTCGTTCCTTAGGCTCTGCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606573 | ||||||||||||||||||||
Literature Links: |
FRK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FRK - fyn related Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002031.2 | 2073 | Missense Mutation | ACA,ATA | T,I 479 | NP_002022.1 | |
XM_005266880.4 | 2073 | Missense Mutation | ACA,ATA | T,I 479 | XP_005266937.1 | |
XM_005266881.2 | 2073 | Missense Mutation | ACA,ATA | T,I 479 | XP_005266938.1 | |
XM_005266882.4 | 2073 | Missense Mutation | ACA,ATA | T,I 479 | XP_005266939.1 | |
XM_011535653.2 | 2073 | Missense Mutation | ACA,ATA | T,I 479 | XP_011533955.1 | |
XM_011535654.2 | 2073 | Missense Mutation | ACA,ATA | T,I 479 | XP_011533956.1 | |
XM_011535655.2 | 2073 | Missense Mutation | ACA,ATA | T,I 479 | XP_011533957.1 | |
XM_011535656.2 | 2073 | Missense Mutation | ACA,ATA | T,I 366 | XP_011533958.1 | |
XM_017010645.1 | 2073 | Missense Mutation | ACA,ATA | T,I 479 | XP_016866134.1 |