Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTGCCAGGTGCTGCGGATGGCA[A/G]TCCTGCTGTCTTACTGCTCTATCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608514 | ||||||||||||||||||||
Literature Links: |
AIG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AIG1 - androgen induced 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286587.1 | 102 | Missense Mutation | ATC,GTC | I,V 3 | NP_001273516.1 | |
NM_001286588.1 | 102 | Missense Mutation | ATC,GTC | I,V 13 | NP_001273517.1 | |
NM_001286589.1 | 102 | Missense Mutation | ATC,GTC | I,V 13 | NP_001273518.1 | |
NM_016108.3 | 102 | Missense Mutation | ATC,GTC | I,V 13 | NP_057192.2 | |
XM_011535881.2 | 102 | Missense Mutation | ATC,GTC | I,V 13 | XP_011534183.1 | |
XM_011535882.2 | 102 | Missense Mutation | ATC,GTC | I,V 13 | XP_011534184.1 | |
XM_011535883.2 | 102 | Missense Mutation | ATC,GTC | I,V 13 | XP_011534185.1 | |
XM_011535884.2 | 102 | Missense Mutation | ATC,GTC | I,V 13 | XP_011534186.1 | |
XM_011535885.2 | 102 | Missense Mutation | ATC,GTC | I,V 13 | XP_011534187.1 | |
XM_017010927.1 | 102 | Missense Mutation | ATC,GTC | I,V 13 | XP_016866416.1 | |
XM_017010928.1 | 102 | Missense Mutation | ATC,GTC | I,V 13 | XP_016866417.1 |