Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACTTCGGCATGGCCTCTTTACAG[A/G]GCACCTTCTGCTCTCAGGTAAGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616805 MIM: 192320 | ||||||||||||||||||||
Literature Links: |
MYCT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYCT1 - myc target 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VIP - vasoactive intestinal peptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003381.3 | 215 | Silent Mutation | AGA,AGG | R,R 30 | NP_003372.1 | |
NM_194435.2 | 215 | Silent Mutation | AGA,AGG | R,R 30 | NP_919416.1 | |
XM_005267135.3 | 215 | Silent Mutation | AGA,AGG | R,R 30 | XP_005267192.1 | |
XM_006715562.3 | 215 | Silent Mutation | AGA,AGG | R,R 30 | XP_006715625.1 |