Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTGGGAGGATCAGCGCCAGCCA[C/T]GCCAGGCCCAGGAGGATCCAGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607369 MIM: 607370 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNK16 - potassium two pore domain channel subfamily K member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135105.1 | 749 | Intron | NP_001128577.1 | |||
NM_001135106.1 | 749 | Silent Mutation | GCA,GCG | A,A 245 | NP_001128578.1 | |
NM_001135107.1 | 749 | Intron | NP_001128579.1 | |||
NM_032115.3 | 749 | Silent Mutation | GCA,GCG | A,A 245 | NP_115491.1 | |
XM_011514935.2 | 749 | Intron | XP_011513237.1 | |||
XM_017011346.1 | 749 | Intron | XP_016866835.1 |
KCNK17 - potassium two pore domain channel subfamily K member 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |