Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTACACGCACTTCAGAATGAAGA[C/G]TTGTAAGTCGCTGACCTGGGGCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603298 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507547 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100507547 - uncharacterized LOC100507547 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPT2 - palmitoyl-protein thioesterase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204103.1 | 183 | Intron | NP_001191032.1 | |||
NM_005155.6 | 183 | Intron | NP_005146.4 | |||
NM_138717.2 | 183 | Missense Mutation | ACT,AGT | T,S 3 | NP_619731.2 |
PPT2-EGFL8 - PPT2-EGFL8 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRRT1 - proline rich transmembrane protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |