Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTACTCTTCCAACGACAGGCCACT[A/G]AAGCTGGTGTCTCTGGGGCGAGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147267 MIM: 614461 | ||||||||||||||||||||
Literature Links: |
ITPR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ITPR3 - inositol 1,4,5-trisphosphate receptor type 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3934 - microRNA 3934 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UQCC2 - ubiquinol-cytochrome c reductase complex assembly factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032340.3 | 305 | Silent Mutation | TTC,TTT | F,F 80 | NP_115716.1 |