Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCATTGGTATCTCTGTCTGTGTC[C/T]GTAACAGTCTGTTTGCCTGTCTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607169 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRSS16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRSS16 - protease, serine 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005865.3 | Intron | NP_005856.1 | ||||
XM_017010161.1 | Intron | XP_016865650.1 | ||||
XM_017010162.1 | Intron | XP_016865651.1 | ||||
XM_017010163.1 | Intron | XP_016865652.1 | ||||
XM_017010164.1 | Intron | XP_016865653.1 | ||||
XM_017010165.1 | Intron | XP_016865654.1 |