Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATAACATTTTATATAACTCATA[A/G]AACAGTGTTTGTATAAAAATTCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616374 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BEND3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BEND3 - BEN domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080450.2 | 6624 | UTR 3 | NP_001073919.1 | |||
XM_005267079.3 | 6624 | Intron | XP_005267136.1 | |||
XM_005267080.3 | 6624 | UTR 3 | XP_005267137.1 | |||
XM_011536005.2 | 6624 | Intron | XP_011534307.1 | |||
XM_017011138.1 | 6624 | UTR 3 | XP_016866627.1 |