Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGTTGTCCACCACTCGTACCCG[C/T]GTCATCTTCTGAATCGCACTCAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611827 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPL14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MRPL14 - mitochondrial ribosomal protein L14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318767.1 | 185 | Silent Mutation | ACA,ACG | T,T 36 | NP_001305696.1 | |
NM_001318768.1 | 185 | Silent Mutation | ACA,ACG | T,T 36 | NP_001305697.1 | |
NM_001318769.1 | 185 | Silent Mutation | ACA,ACG | T,T 47 | NP_001305698.1 | |
NM_001318770.1 | 185 | Silent Mutation | ACA,ACG | T,T 89 | NP_001305699.1 | |
NM_001318771.1 | 185 | Silent Mutation | ACA,ACG | T,T 2 | NP_001305700.1 | |
NM_032111.3 | 185 | Silent Mutation | ACA,ACG | T,T 36 | NP_115487.2 |