Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGTGAGCACCAACTGCGGACAT[C/G]TCTTCTGTCGAGTGTGCCTGACACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616976 | ||||||||||||||||||||
Literature Links: |
TRIM40 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM40 - tripartite motif containing 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286633.1 | 911 | Missense Mutation | CTC,GTC | L,V 32 | NP_001273562.1 | |
NM_138700.4 | 911 | Missense Mutation | CTC,GTC | L,V 32 | NP_619645.1 | |
XM_011514305.1 | 911 | Missense Mutation | CTC,GTC | L,V 32 | XP_011512607.1 | |
XM_011514306.1 | 911 | Missense Mutation | CTC,GTC | L,V 32 | XP_011512608.1 | |
XM_011514308.1 | 911 | Missense Mutation | CTC,GTC | L,V 32 | XP_011512610.1 | |
XM_011514309.1 | 911 | Missense Mutation | CTC,GTC | L,V 32 | XP_011512611.1 |