Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTTTCCTTTCAGGAGCCCCCCA[A/G]GGTTGCCAAATGCACAGCCAAGCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 143170 MIM: 601646 | ||||||||||||||||||||
Literature Links: |
MEA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MEA1 - male-enhanced antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318942.1 | 217 | Intron | NP_001305871.1 | |||
NM_001318943.1 | 217 | Intron | NP_001305872.1 | |||
NM_014623.3 | 217 | Intron | NP_055438.1 | |||
XM_005249121.3 | 217 | Intron | XP_005249178.1 | |||
XM_017010868.1 | 217 | Intron | XP_016866357.1 |
PPP2R5D - protein phosphatase 2 regulatory subunit B'delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270476.1 | 217 | UTR 5 | NP_001257405.1 | |||
NM_006245.3 | 217 | Missense Mutation | AAG,AGG | K,R 13 | NP_006236.1 | |
NM_180976.2 | 217 | Missense Mutation | AAG,AGG | K,R 13 | NP_851307.1 | |
NM_180977.2 | 217 | Intron | NP_851308.1 |