Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAGATGGCGGCCTCCTGGTCGC[C/T]CTTGGTTACCCTGCGCCCCTTAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LACE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LACE1 - lactation elevated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323005.1 | 109 | Missense Mutation | CCC,CTC | P,L 7 | NP_001309934.1 | |
NM_145315.4 | 109 | Missense Mutation | CCC,CTC | P,L 7 | NP_660358.2 | |
XM_005266885.3 | 109 | Intron | XP_005266942.1 | |||
XM_011535657.2 | 109 | Missense Mutation | CCC,CTC | P,L 7 | XP_011533959.1 | |
XM_011535659.2 | 109 | Missense Mutation | CCC,CTC | P,L 7 | XP_011533961.1 | |
XM_011535660.2 | 109 | Missense Mutation | CCC,CTC | P,L 7 | XP_011533962.1 | |
XM_011535661.2 | 109 | Missense Mutation | CCC,CTC | P,L 7 | XP_011533963.1 | |
XM_017010647.1 | 109 | Intron | XP_016866136.1 | |||
XM_017010648.1 | 109 | Intron | XP_016866137.1 | |||
XM_017010649.1 | 109 | UTR 5 | XP_016866138.1 |