Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGAGACTAGCGAGTAATGAAGTA[A/C]TAATACAGAGTCAGCCCCCATTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607995 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UNC93A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UNC93A - unc-93 homolog A (C. elegans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143947.1 | Intron | NP_001137419.1 | ||||
NM_018974.3 | Intron | NP_061847.2 | ||||
XM_011535905.2 | Intron | XP_011534207.1 | ||||
XM_011535906.2 | Intron | XP_011534208.1 | ||||
XM_011535907.2 | Intron | XP_011534209.1 | ||||
XM_011535908.2 | Intron | XP_011534210.1 | ||||
XM_017010958.1 | Intron | XP_016866447.1 |