Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGTTTCAGGAGAGCTCTGCCCA[A/G]GGCCTGGGCCTGAGCCTGGGGCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607169 | ||||||||||||||||||||
Literature Links: |
PRSS16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRSS16 - protease, serine 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005865.3 | 144 | Silent Mutation | CAA,CAG | Q,Q 43 | NP_005856.1 | |
XM_017010161.1 | 144 | Silent Mutation | CAA,CAG | Q,Q 43 | XP_016865650.1 | |
XM_017010162.1 | 144 | Silent Mutation | CAA,CAG | Q,Q 43 | XP_016865651.1 | |
XM_017010163.1 | 144 | Silent Mutation | CAA,CAG | Q,Q 43 | XP_016865652.1 | |
XM_017010164.1 | 144 | Silent Mutation | CAA,CAG | Q,Q 43 | XP_016865653.1 | |
XM_017010165.1 | 144 | Silent Mutation | CAA,CAG | Q,Q 43 | XP_016865654.1 |