Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCGGGCATCAACATGCAGAAA[C/G]TTGCGATGCCTGCTGTGGCAGCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 130135 | ||||||||||||||||||||
Literature Links: |
SERPINB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERPINB1 - serpin family B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030666.3 | 1405 | Missense Mutation | ACT,AGT | T,S 342 | NP_109591.1 | |
XM_011514333.1 | 1405 | Missense Mutation | ACT,AGT | T,S 342 | XP_011512635.1 | |
XM_011514334.2 | 1405 | Missense Mutation | ACT,AGT | T,S 342 | XP_011512636.1 | |
XM_011514335.2 | 1405 | Missense Mutation | ACT,AGT | T,S 293 | XP_011512637.1 |