Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGCGGCCGCAGAAGAGAATCCC[A/G]TTGGTCTTGCTGTGCTGGATGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 173321 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINB6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERPINB6 - serpin family B member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195291.2 | 1679 | Silent Mutation | AAC,AAT | N,N 369 | NP_001182220.2 | |
NM_001271822.1 | 1679 | Silent Mutation | AAC,AAT | N,N 379 | NP_001258751.1 | |
NM_001271823.1 | 1679 | Silent Mutation | AAC,AAT | N,N 384 | NP_001258752.1 | |
NM_001271824.1 | 1679 | Silent Mutation | AAC,AAT | N,N 365 | NP_001258753.1 | |
NM_001271825.1 | 1679 | Silent Mutation | AAC,AAT | N,N 365 | NP_001258754.1 | |
NM_001297699.1 | 1679 | Silent Mutation | AAC,AAT | N,N 365 | NP_001284628.1 | |
NM_001297700.1 | 1679 | Silent Mutation | AAC,AAT | N,N 365 | NP_001284629.1 | |
NM_004568.5 | 1679 | Silent Mutation | AAC,AAT | N,N 365 | NP_004559.4 | |
XM_011514672.1 | 1679 | Silent Mutation | AAC,AAT | N,N 443 | XP_011512974.1 | |
XM_011514673.1 | 1679 | Silent Mutation | AAC,AAT | N,N 365 | XP_011512975.1 | |
XM_011514674.2 | 1679 | Silent Mutation | AAC,AAT | N,N 365 | XP_011512976.1 | |
XM_011514676.2 | 1679 | Silent Mutation | AAC,AAT | N,N 321 | XP_011512978.1 | |
XM_017010940.1 | 1679 | Silent Mutation | AAC,AAT | N,N 369 | XP_016866429.1 | |
XM_017010941.1 | 1679 | Silent Mutation | AAC,AAT | N,N 321 | XP_016866430.1 |